missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-fluorobenzaldehyde, 98%, Thermo Scientific Chemicals

Catalog Number p-7021753
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
93777-26-5
C7H4BrFO
203.01
MFCD00070755
MMFGGDVQLQQQRX-UHFFFAOYSA-N
2-fluoro-5-bromobenzaldehyde, 5-bromo-2-fluoro-benzaldehyde, 2-fluoro-5-bromo benzaldehyde, 5-bromo-2-fluoro benzaldehyde, benzaldehyde, 5-bromo-2-fluoro, 5-brom-2-fluorbenzaldehyd, pubchem2879, acmc-209rnk, 3-bromo-6-fluorobenzaldehyde, ksc494e9t
736327
5-bromo-2-fluorobenzaldehyde
C1=CC(=C(C=C1Br)C=O)F
This item is not returnable. View return policy
93777-26-5
C7H4BrFO
203.01
MFCD00070755
MMFGGDVQLQQQRX-UHFFFAOYSA-N
2-fluoro-5-bromobenzaldehyde, 5-bromo-2-fluoro-benzaldehyde, 2-fluoro-5-bromo benzaldehyde, 5-bromo-2-fluoro benzaldehyde, benzaldehyde, 5-bromo-2-fluoro, 5-brom-2-fluorbenzaldehyd, pubchem2879, acmc-209rnk, 3-bromo-6-fluorobenzaldehyde, ksc494e9t
736327
5-bromo-2-fluorobenzaldehyde
C1=CC(=C(C=C1Br)C=O)F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.71
Boiling Point 230°C
Flash Point 109°C (228°F)
Refractive Index 1.57
Quantity 10 g
Beilstein 7632798
Sensitivity Air Sensitive
Formula Weight 203.01
Percent Purity 98%
Chemical Name or Material 5-Bromo-2-fluorobenzaldehyde

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'