missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5'-Bromo-2'-fluoroacetophenone, 98%, Thermo Scientific Chemicals
47.80 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
Description
5'-Bromo-2'-fluoroacetophenone is an intermediate used in pharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5′-Bromo-2′-fluoroacetophenone is an intermediate used in pharmaceuticals.
Solubility
Slightly soluble in water.
Notes
Recommended storage temp 2-8°C. Store in cool, well ventilated area. Keep container tightly closed.
5′-Bromo-2′-fluoroacetophenone is an intermediate used in pharmaceuticals.
Solubility
Slightly soluble in water.
Notes
Recommended storage temp 2-8°C. Store in cool, well ventilated area. Keep container tightly closed.
Specifications
Quantity | 5 g |
Solubility Information | Slightly soluble in water. |
Formula Weight | 217.04 |
Percent Purity | 98% |
Physical Form | Solid |
Chemical Name or Material | 5'-Bromo-2'-fluoroacetophenone |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'