missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromo-2-chloropyrimidine, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
It is applied as a pharmaceutical intermediate, in the synthesis of pharmaceutical goods such as inhibitors. It is also employed in the cross-coupling reactions of indium organometallics with 2,5-dihalopyrimidines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is applied as a pharmaceutical intermediate, in the synthesis of pharmaceutical goods such as inhibitors. It is also employed in the cross-coupling reactions of indium organometallics with 2,5-dihalopyrimidines.
Solubility
Insoluble in water. Slightly soluble in DMSO and methanol.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
It is applied as a pharmaceutical intermediate, in the synthesis of pharmaceutical goods such as inhibitors. It is also employed in the cross-coupling reactions of indium organometallics with 2,5-dihalopyrimidines.
Solubility
Insoluble in water. Slightly soluble in DMSO and methanol.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Melting Point | 77°C to 80°C |
Boiling Point | 95°C (15 mmHg) |
Quantity | 5 g |
Beilstein | 507955 |
Solubility Information | Insoluble in water. Slightly soluble in DMSO and methanol. |
Formula Weight | 193.43 |
Percent Purity | ≥98% |
Chemical Name or Material | 5-Bromo-2-chloropyrimidine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'