missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-chloropyrimidine, 96%, Thermo Scientific Chemicals

Catalog Number p-3777617
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
32779-36-5
C4H2BrClN2
193.43
MFCD00483232
XPGIBDJXEVAVTO-UHFFFAOYSA-N
2-chloro-5-bromopyrimidine, 5-bromo-2-chloro-pyrimidine, 5-bromo-2-chloropyrimidne, pyrimidine, 5-bromo-2-chloro, 5-bromo-2-chloro-1,3-diazine, pubchem5159, zlchem 1210, acmc-209hvm, 5-bromo chloropyrimidine, 5-bromo-2-chloroprimidine
606665
5-bromo-2-chloropyrimidine
ClC1=NC=C(Br)C=N1
This item is not returnable. View return policy
32779-36-5
C4H2BrClN2
193.43
MFCD00483232
XPGIBDJXEVAVTO-UHFFFAOYSA-N
2-chloro-5-bromopyrimidine, 5-bromo-2-chloro-pyrimidine, 5-bromo-2-chloropyrimidne, pyrimidine, 5-bromo-2-chloro, 5-bromo-2-chloro-1,3-diazine, pubchem5159, zlchem 1210, acmc-209hvm, 5-bromo chloropyrimidine, 5-bromo-2-chloroprimidine
606665
5-bromo-2-chloropyrimidine
ClC1=NC=C(Br)C=N1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 95.0
CAS Max % 100.0
Melting Point 78.0°C to 80.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 95% min. (HPLC)
Packaging Glass bottle
Quantity 250 mg
Solubility Information Solubility in water: insoluble in water
Formula Weight 193.43
Percent Purity 96%
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-chloropyrimidine
Show More Show Less
compliance-icons
Product Identifier
  • 5-Bromo-2-chloropyrimidine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'