missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Bromo-2-chlorobenzaldehyde, 98%

Brand:  Alfa Aesar™ H64438.03

Code : NEW

Additional Details : CAS Number : 189628-37-3 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15461649

  • £32.40 / 1g


Estimated Shipment date
from Supplier 23-02-2018
Add to basket

Description

Description

Applications
It finds its application as an intermediate in organic syntheses.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Chemical Identifiers

5-Bromo-2-chlorobenzaldehyde
219.46
9757388
189628-37-3
MFCD08445659

Specifications

5-Bromo-2-chlorobenzaldehyde
Off-white to pale yellow
Solid
219.46
9757388
189628-37-3
1g
98%
MFCD08445659
Videos
SDS