missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Bromo-2-chloro-4-methyl-3-nitropyridine, 98%

Brand:  Alfa Aesar™ H61157.06

Code : NEW

Additional Details : CAS Number : 884495-15-2 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15427668

  • £358.49 / 5g


Estimated Shipment date
from Supplier 05-03-2018
Add to basket

Specifications

Chemical Identifiers

5-Bromo-2-chloro-4-methyl-3-nitropyridine
884495-15-2
251.47
MFCD06659505

Specifications

5-Bromo-2-chloro-4-methyl-3-nitropyridine
Solid
251.47
5g
884495-15-2
98%
MFCD06659505
Videos
SDS