missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-chloro-3-nitropyridine, 98%, Thermo Scientific Chemicals

Catalog Number p-7056136
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
67443-38-3
C5H2BrClN2O2
237.44
MFCD00222270
WWQQPSDIIVXFOX-UHFFFAOYSA-N
2-chloro-3-nitro-5-bromopyridine, 5-bromo-2-chloro-3-nitro-pyridine, pyridine, 5-bromo-2-chloro-3-nitro, abbypharma ap-12-5159, zlchem 393, pubchem1166, acmc-209nz1, ksc352q2l, 5-bromo-2-chloro-3-nitropyridin, 2-chloro-5-bromo-3-nitropyridine
7019412
5-bromo-2-chloro-3-nitropyridine
[O-][N+](=O)C1=CC(Br)=CN=C1Cl
This item is not returnable. View return policy
67443-38-3
C5H2BrClN2O2
237.44
MFCD00222270
WWQQPSDIIVXFOX-UHFFFAOYSA-N
2-chloro-3-nitro-5-bromopyridine, 5-bromo-2-chloro-3-nitro-pyridine, pyridine, 5-bromo-2-chloro-3-nitro, abbypharma ap-12-5159, zlchem 393, pubchem1166, acmc-209nz1, ksc352q2l, 5-bromo-2-chloro-3-nitropyridin, 2-chloro-5-bromo-3-nitropyridine
7019412
5-bromo-2-chloro-3-nitropyridine
[O-][N+](=O)C1=CC(Br)=CN=C1Cl

5-Bromo-2-chloro-3-nitropyridine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromo-2-chloro-3-nitropyridine is used as pharmaceutical intermediate.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 66°C to 68°C
Color Yellow
Quantity 25 g
UN Number UN2811
Beilstein 147551
Formula Weight 237.44
Percent Purity 98%
Physical Form Crystalline
Chemical Name or Material 5-Bromo-2-chloro-3-nitropyridine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'