missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Bromo-2-chloro-3-nitropyridine, 98%

Brand:  Alfa Aesar™ H64897.22

Code : NEW

Additional Details : CAS Number : 67443-38-3 Weight : 0.10000kg Transport : UN number : 2811 Chem class : 1 Pack group : III

Quantity 100g

 View more versions of this product

Product Code. 15452299

  • £126.00 / 100g


Estimated Shipment date
from Supplier 24-01-2018
Add to basket

Description

Description

Applications
5-Bromo-2-chloro-3-nitropyridine is used as pharmaceutical intermediate.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

5-Bromo-2-chloro-3-nitropyridine
237.44
UN2811
67443-38-3
MFCD00222270
147551

Specifications

5-Bromo-2-chloro-3-nitropyridine
Crystalline
237.44
UN2811
Yellow
100g
67443-38-3
98%
MFCD00222270
147551
66° to 68°C
Videos
SDS