missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Bromo-2-chloro-3-cyanopyridine, 98%

Brand:  Alfa Aesar™ H61664.06

Code : NEW

Additional Details : CAS Number : 405224-23-9 Weight : 0.00500kg Transport : UN number : 3439 Chem class : 6.1 Pack group : III

Quantity 5g

 View more versions of this product

Product Code. 15408228

  • £117.90 / 5g


Estimated Shipment date
from Supplier 19-06-2018
Add to basket

Description

Description

Applications
5-Bromo-2-chloro-3-cyanopyridine is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

5-Bromo-2-chloro-3-cyanopyridine
217.45
UN3439
405224-23-9
MFCD09801046
5-Bromo-2-chloronicotinonitrile; 5-Bromo-2-chloropyridine-3-carbonitrile

Specifications

5-Bromo-2-chloro-3-cyanopyridine
5g
98%
MFCD09801046
5-Bromo-2-chloronicotinonitrile; 5-Bromo-2-chloropyridine-3-carbonitrile
405224-23-9
Solid
217.45
UN3439
Videos
SDS