missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-anisaldehyde, 99%, ACROS Organics™

Brand:  Acros Organics 167520250

Code : 29

Additional Details : CAS Number : 25016-01-7 Weight : 0.07500kg


Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10701061

  • £31.90 / 25g


Estimated Shipment
11-05-2018
Add to basket


Specifications

Chemical Identifiers

5-Bromo-2-anisaldehyde
98.5
≥98.5%
C8H7BrO2
215.05
5-Bromo-2-methoxybenzaldehyde
25016-01-7
100.0
99%
BrC6H3(OCH3)CHO
MFCD00003347

Specifications

5-Bromo-2-anisaldehyde
98.5
Authentic
Crystalline Powder
99%
BrC6H3(OCH3)CHO
MFCD00003347
08, 55
5-Bromo-2-methoxybenzaldehyde
25016-01-7
100.0
Glass bottle
≥98.5%
C8H7BrO2
215.05
115° to 118°C
Light yellow
25g
Videos
SDS