missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2,4-dichloropyrimidine, 98%, Thermo Scientific Chemicals

Catalog Number p-3777619
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
36082-50-5
C4HBrCl2N2
227.88
MFCD00127818
SIKXIUWKPGWBBF-UHFFFAOYSA-N
2,4-dichloro-5-bromopyrimidine, 2,4-dichloro-5-bromo pyrimidine, 5-bromo-2,4-dichloro-pyrimidine, pyrimidine, 5-bromo-2,4-dichloro, 5-bromo-2,6-dichloropyrimidine, 5-bromo-2,4-dichloro-1,3-diazine, 5-brom-2,4-dichlorpyrimidin, zlchem 380, pubchem3993, acmc-1ad68
289973
5-bromo-2,4-dichloropyrimidine
C1=C(C(=NC(=N1)Cl)Cl)Br
This item is not returnable. View return policy
36082-50-5
C4HBrCl2N2
227.88
MFCD00127818
SIKXIUWKPGWBBF-UHFFFAOYSA-N
2,4-dichloro-5-bromopyrimidine, 2,4-dichloro-5-bromo pyrimidine, 5-bromo-2,4-dichloro-pyrimidine, pyrimidine, 5-bromo-2,4-dichloro, 5-bromo-2,6-dichloropyrimidine, 5-bromo-2,4-dichloro-1,3-diazine, 5-brom-2,4-dichlorpyrimidin, zlchem 380, pubchem3993, acmc-1ad68
289973
5-bromo-2,4-dichloropyrimidine
C1=C(C(=NC(=N1)Cl)Cl)Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 29.0°C to 30.0°C
Color Colorless to Yellow
Density 1.7810g/mL
Boiling Point 128.0°C (15.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.6020 to 1.6040 (20°C, 589nm)
Quantity 1 g
Specific Gravity 1.781
Formula Weight 227.88
Percent Purity 98%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material 5-Bromo-2, 4-dichloropyrimidine
Show More Show Less
compliance-icons
Product Identifier
  • 5-Bromo-2,4-dichloropyrimidine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'