missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2,3-dihydrobenzo[b]furan-7-carbaldehyde, 97%, Thermo Scientific™

Catalog Number p-4510233
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
281678-73-7
C9H7BrO2
227.057
LQCBHNHULMHKFB-UHFFFAOYSA-N
5-bromo-2,3-dihydrobenzo b furan-7-carbaldehyde, 5-bromo-2,3-dihydrobenzofuran-7-carbaldehyde, 5-bromo-2,3-dihydobenzo b furan-7-aldehyde, 5-bromo-2,3-dihydrobenzofuran 7-carbaldehyde, 5-bromo-2,3-dihydrobenzo b furane-7-carbaldehyde, 7-benzofurancarboxaldehyde, 5-bromo-2,3-dihydro
2794804
5-bromo-2,3-dihydro-1-benzofuran-7-carbaldehyde
C1COC2=C1C=C(C=C2C=O)Br
This item is not returnable. View return policy
281678-73-7
C9H7BrO2
227.057
LQCBHNHULMHKFB-UHFFFAOYSA-N
5-bromo-2,3-dihydrobenzo b furan-7-carbaldehyde, 5-bromo-2,3-dihydrobenzofuran-7-carbaldehyde, 5-bromo-2,3-dihydobenzo b furan-7-aldehyde, 5-bromo-2,3-dihydrobenzofuran 7-carbaldehyde, 5-bromo-2,3-dihydrobenzo b furane-7-carbaldehyde, 7-benzofurancarboxaldehyde, 5-bromo-2,3-dihydro
2794804
5-bromo-2,3-dihydro-1-benzofuran-7-carbaldehyde
C1COC2=C1C=C(C=C2C=O)Br
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 227.06
Percent Purity 97%
Chemical Name or Material 5-Bromo-2, 3-dihydrobenzo[b]furan-7-carbaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'