missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2,3-dihydrobenzo[b]furan-7-boronic acid, 97%, Thermo Scientific™

Catalog Number p-3075763
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
690632-72-5
C8H8BBrO3
242.86
MFCD06200893
BSSWVOKZPQTKGW-UHFFFAOYSA-N
5-bromo-2,3-dihydrobenzofuran-7-yl boronic acid, 5-bromo-2,3-dihydrobenzo b furan-7-boronic acid, 5-bromo-2,3-dihydro-1-benzofuran-7-yl boronic acid, acmc-1b7su, 5-bromo-2,3-dihydrobenzofuran-7-yl boronicacid, boronic acid, b-5-bromo-2,3-dihydro-7-benzofuranyl
2794805
(5-bromo-2,3-dihydro-1-benzofuran-7-yl)boronic acid
OB(O)C1=CC(Br)=CC2=C1OCC2
This item is not returnable. View return policy
690632-72-5
C8H8BBrO3
242.86
MFCD06200893
BSSWVOKZPQTKGW-UHFFFAOYSA-N
5-bromo-2,3-dihydrobenzofuran-7-yl boronic acid, 5-bromo-2,3-dihydrobenzo b furan-7-boronic acid, 5-bromo-2,3-dihydro-1-benzofuran-7-yl boronic acid, acmc-1b7su, 5-bromo-2,3-dihydrobenzofuran-7-yl boronicacid, boronic acid, b-5-bromo-2,3-dihydro-7-benzofuranyl
2794805
(5-bromo-2,3-dihydro-1-benzofuran-7-yl)boronic acid
OB(O)C1=CC(Br)=CC2=C1OCC2
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 242.86
Percent Purity 97%
Chemical Name or Material 5-Bromo-2, 3-dihydrobenzo[b]furan-7-boronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'