missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromo-2,3-difluoropyridine, 97%, Thermo Scientific Chemicals
279.34 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
Description
It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications
Color | Colorless to Yellow |
Quantity | 25 g |
Beilstein | 13412561 |
Synonym | pyridine, 5-bromo-2,3-difluoro, 2,3-difluoro-5-bromopyridine, pubchem1292, 5-bromo-2,3-difluoro-pyridine, pyridine,5-bromo-2,3-difluoro, 5-bromanyl-2,3-bis fluoranyl pyridine |
Solubility Information | Slightly soluble in water. |
Formula Weight | 193.98 |
Percent Purity | 97% |
Physical Form | Liquid |
Chemical Name or Material | 5-Bromo-2,3-difluoropyridine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'