missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-bromo-1h-indazole, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Specifications
Assay Percent Range | 97% |
Quantity | 1 g |
Formula Weight | 197.03 |
Percent Purity | 97% |
Chemical Name or Material | 5-Bromo-1H-indazole |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'