missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromo-1H-indazole-3-carboxylic acid, 95%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Specifications
Assay Percent Range | 95% |
Quantity | 250 mg |
Synonym | 5-bromo-1h-indazole-3-carboxylicacid, 5-bromoindazole-3-carboxylic acid, 5-bromo-3-indazolecarboxylic acid, 1h-indazole-3-carboxylic acid, 5-bromo, 1h-indazole-3-carboxylic acid,5-bromo, pubchem7817, acmc-2098xf, ksc173q5b, 5-bromo-3-carboxy-1h-indazole, 1h-indazole-3-carboxylicacid, 5-bromo |
Formula Weight | 241.04 |
Percent Purity | 95% |
Chemical Name or Material | 5-Bromo-1H-indazole-3-carboxylic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'