missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-1-methylindoline, 90%, Thermo Scientific™

Catalog Number p-3586805
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
99848-78-9
C9H10BrN
212.09
MFCD09817498
YIRSVRZXIXHOSN-UHFFFAOYSA-N
5-bromo-1-methylindoline, 5-bromo-1-methyl-2,3-dihydro-1h-indole, 1h-indole, 5-bromo-2,3-dihydro-1-methyl, 1-methyl-5-bromo-2,3-dihydro-1h-indole, 5-bromo-2,3-dihydro-1-methyl-1h-indole
19361435
CN1CCC2=C1C=CC(Br)=C2
This item is not returnable. View return policy
99848-78-9
C9H10BrN
212.09
MFCD09817498
YIRSVRZXIXHOSN-UHFFFAOYSA-N
5-bromo-1-methylindoline, 5-bromo-1-methyl-2,3-dihydro-1h-indole, 1h-indole, 5-bromo-2,3-dihydro-1-methyl, 1-methyl-5-bromo-2,3-dihydro-1h-indole, 5-bromo-2,3-dihydro-1-methyl-1h-indole
19361435
CN1CCC2=C1C=CC(Br)=C2
CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 250 mg
IUPAC Name 5-bromo-1-methyl-2,3-dihydroindole
Formula Weight 212.09
Percent Purity 90%
Chemical Name or Material 5-Bromo-1-methylindoline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'