missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-1-methyl-2-oxoindoline, 97%, Thermo Scientific™

Catalog Number p-3522549
Click to view available options
Quantity:
1 g
5 g
10 g
This item is not returnable. View return policy
20870-90-0
C9H8BrNO
226.07
MFCD00464061
WARSUKBSFLACOI-UHFFFAOYSA-N
5-bromo-1-methyl-2-oxoindoline, 5-bromo-1-methylindolin-2-one, 5-bromo-1-methyl-1,3-dihydro-indol-2-one, 5-bromo-1-methyl-2,3-dihydro-1h-indol-2-one, 5-bromo-1-methyl-1,3-dihydro-2h-indol-2-one, 5-bromo-1-methyl-1h-indolin-2-one, acmc-209ffi, 5-bromo-1-methyl oxindole, 1-methyl-5-bromoindoline-2-one
2432666
5-bromo-1-methyl-3H-indol-2-one
CN1C(=O)CC2=CC(Br)=CC=C12
This item is not returnable. View return policy
20870-90-0
C9H8BrNO
226.07
MFCD00464061
WARSUKBSFLACOI-UHFFFAOYSA-N
5-bromo-1-methyl-2-oxoindoline, 5-bromo-1-methylindolin-2-one, 5-bromo-1-methyl-1,3-dihydro-indol-2-one, 5-bromo-1-methyl-2,3-dihydro-1h-indol-2-one, 5-bromo-1-methyl-1,3-dihydro-2h-indol-2-one, 5-bromo-1-methyl-1h-indolin-2-one, acmc-209ffi, 5-bromo-1-methyl oxindole, 1-methyl-5-bromoindoline-2-one
2432666
5-bromo-1-methyl-3H-indol-2-one
CN1C(=O)CC2=CC(Br)=CC=C12
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 226.07
Percent Purity 97%
Chemical Name or Material 5-Bromo-1-methyl-2-oxoindoline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'