missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-1-benzofuran, 97%, Thermo Scientific™

Catalog Number 10641673
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
23145-07-5
C8H5BrO
197.031
MFCD03407317
AYOVPQORFBWFNO-UHFFFAOYSA-N
5-bromobenzofuran, benzofuran, 5-bromo, 5-bromobenzo b furan, 5-bromo-benzofuran, 5-bromobenzo beta furan, buttpark 95\50-66, pubchem22017, 5-bromobenzo b-furan, #
90015
5-bromo-1-benzofuran
C1=CC2=C(C=CO2)C=C1Br
This item is not returnable. View return policy
23145-07-5
C8H5BrO
197.031
MFCD03407317
AYOVPQORFBWFNO-UHFFFAOYSA-N
5-bromobenzofuran, benzofuran, 5-bromo, 5-bromobenzo b furan, 5-bromo-benzofuran, 5-bromobenzo beta furan, buttpark 95\50-66, pubchem22017, 5-bromobenzo b-furan, #
90015
5-bromo-1-benzofuran
C1=CC2=C(C=CO2)C=C1Br
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 197.03
Percent Purity 97%
Chemical Name or Material 5-Bromo-1-benzofuran
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'