missing translation for 'onlineSavingsMsg'
Learn More

5-Benzyloxy-3-fluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific™

Catalog Number 15407626
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1528757-88-1
C19H22BFO3
328.19
FAVUCVNUXHFOPD-UHFFFAOYSA-N
3-Benzyloxy-5-fluorophenylboronic acid pinacol ester
81315028
2-(3-fluoro-5-phenylmethoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC(=C2)F)OCC3=CC=CC=C3
This item is not returnable. View return policy
1528757-88-1
C19H22BFO3
328.19
FAVUCVNUXHFOPD-UHFFFAOYSA-N
3-Benzyloxy-5-fluorophenylboronic acid pinacol ester
81315028
2-(3-fluoro-5-phenylmethoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC(=C2)F)OCC3=CC=CC=C3

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 328.19
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 5-Benzyloxy-3-fluorobenzeneboronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'