Learn More
Tocris Bioscience™ 5-BDBD
Potent and selective P2X4 antagonist
Brand:  Tocris Bioscience™ 3579/10
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13443234
  • £198.32 / 10mg
Estimated Shipment
11-09-2020
Description
Description
Potent and selective P2X4 receptor antagonist. Blocks P2X4-mediated currents in Chinese hamster ovary cells (IC50 = 0.50 μM). Exhibits no significant antagonist effects at other P2X receptors. Reduces long-term potentiation (LTP) in rat hippocampal slices.
Specifications
Specifications
5-BDBD
>99
10mg
5-BDBD
355.19