missing translation for 'onlineSavingsMsg'
Learn More

5-Aminonicotinic acid, 95%, Thermo Scientific™

Catalog Number p-3735486
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
24242-19-1
C6H6N2O2
138.13
MFCD00129116
BYIORJAACCWFPU-UHFFFAOYSA-N
5-aminonicotinic acid, 5-amino-nicotinic acid, 5-aminonicotinicacid, 3-pyridinecarboxylic acid, 5-amino, 5-amino nicotinic acid, 5-amino-3-carboxypyridine, 5-amino-3-pyridinecarboxylic acid, zlchem 307, pubchem15914, acmc-209ga2
354316
CHEBI:68578
5-aminopyridine-3-carboxylic acid
C1=C(C=NC=C1N)C(=O)O
This item is not returnable. View return policy
24242-19-1
C6H6N2O2
138.13
MFCD00129116
BYIORJAACCWFPU-UHFFFAOYSA-N
5-aminonicotinic acid, 5-amino-nicotinic acid, 5-aminonicotinicacid, 3-pyridinecarboxylic acid, 5-amino, 5-amino nicotinic acid, 5-amino-3-carboxypyridine, 5-amino-3-pyridinecarboxylic acid, zlchem 307, pubchem15914, acmc-209ga2
354316
CHEBI:68578
5-aminopyridine-3-carboxylic acid
C1=C(C=NC=C1N)C(=O)O
CAS Min % 94.0
CAS Max % 100.0
Color Brown
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 138.13
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 5-Aminonicotinic acid
Show More Show Less
compliance-icons
Product Identifier
  • 5-Aminonicotinic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'