missing translation for 'onlineSavingsMsg'
Learn More

5-Aminonicotinic acid, 95%, ACROS Organics™

Brand:  Acros Organics 361140010

Code : 29

Additional Details : CAS Number : 24242-19-1 Weight : 0.05100kg Transport : UN number : 2811 Chem class : 1 Pack group : II


Quantity 1g
Packaging Glass bottle

 View more versions of this product

Product Code. 10562992

  • £66.40 / 1g


Estimated Shipment
14-03-2018
Add to basket


Specifications

Chemical Identifiers

5-Aminonicotinic acid
94.0
≥94%
C6H6N2O2
MFCD00129116
24242-19-1
100.0
95%
138.13

Specifications

5-Aminonicotinic acid
94.0
Authentic
Golden-brown
≥94%
1g
138.13
24242-19-1
100.0
Glass bottle
Crystalline Powder
95%
C6H6N2O2
MFCD00129116
Videos
SDS