missing translation for 'onlineSavingsMsg'
Learn More

5-Aminoisoquinoline, 99%, Thermo Scientific Chemicals

Catalog Number 10563131
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1125-60-6
C9H8N2
144.18
MFCD00006907
DTVYNUOOZIKEEX-UHFFFAOYSA-N
5-aminoisoquinoline, 5-isoquinolinamine, isoquinoline, 5-amino, isoquinol-5-ylamine, isoquinolin-5-ylamine, 5-amino-isoquinoline, 5-aminoisoquinolin, 5-isoquinolylamine, 5iq, 5aminoisoquinoline
70766
isoquinolin-5-amine
NC1=C2C=CN=CC2=CC=C1
This item is not returnable. View return policy
1125-60-6
C9H8N2
144.18
MFCD00006907
DTVYNUOOZIKEEX-UHFFFAOYSA-N
5-aminoisoquinoline, 5-isoquinolinamine, isoquinoline, 5-amino, isoquinol-5-ylamine, isoquinolin-5-ylamine, 5-amino-isoquinoline, 5-aminoisoquinolin, 5-isoquinolylamine, 5iq, 5aminoisoquinoline
70766
isoquinolin-5-amine
NC1=C2C=CN=CC2=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 127°C to 130°C
Color Brown-Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Beilstein 22, 452
Formula Weight 144.18
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 5-Aminoisoquinoline, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 5-Aminoisoquinoline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'