missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2-(trifluoromethyl)benzimidazole, 96%, Thermo Scientific™

Catalog Number p-4813183
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
3671-66-7
C8H6F3N3
201.15
CKEKFQLHCAZGSP-UHFFFAOYSA-N
5-amino-2-trifluoromethyl benzimidazole, 1h-benzimidazol-5-amine, 2-trifluoromethyl, 2-trifluoromethyl-1h-benzo d imidazol-5-amine, benzimidazole, 5-amino-2-trifluoromethyl, 2-trifluoromethyl-1h-benzimidazol-5-amine, 2-trifluoromethyl-1h-1,3-benzodiazol-5-amine, 5-amino-2-trifluoromethylbenzimidazole, 2-trifluoromethyl benzimidazole-5-ylamine, 2-trifluoromethyl-1h-benzimidazol-6-amine, 2-trifluoromethyl-3h-benzoimidazol-5-ylamine
19345
2-(trifluoromethyl)-3H-benzimidazol-5-amine
C1=CC2=C(C=C1N)NC(=N2)C(F)(F)F
This item is not returnable. View return policy
3671-66-7
C8H6F3N3
201.15
CKEKFQLHCAZGSP-UHFFFAOYSA-N
5-amino-2-trifluoromethyl benzimidazole, 1h-benzimidazol-5-amine, 2-trifluoromethyl, 2-trifluoromethyl-1h-benzo d imidazol-5-amine, benzimidazole, 5-amino-2-trifluoromethyl, 2-trifluoromethyl-1h-benzimidazol-5-amine, 2-trifluoromethyl-1h-1,3-benzodiazol-5-amine, 5-amino-2-trifluoromethylbenzimidazole, 2-trifluoromethyl benzimidazole-5-ylamine, 2-trifluoromethyl-1h-benzimidazol-6-amine, 2-trifluoromethyl-3h-benzoimidazol-5-ylamine
19345
2-(trifluoromethyl)-3H-benzimidazol-5-amine
C1=CC2=C(C=C1N)NC(=N2)C(F)(F)F
Melting Point 189.0°C to 191.0°C
Assay Percent Range 95% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 201.15
Percent Purity 96%
Chemical Name or Material 5-Amino-2-(trifluoromethyl)benzimidazole
compliance-icons
Product Identifier
  • 5-Amino-2-(trifluoromethyl)benzimidazole
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'