missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2-methylbenzonitrile, 97%, ACROS Organics™

Brand:  Acros Organics 439850250

Code : 29

Additional Details : CAS Number : 50670-64-9 Weight : 0.07500kg Transport : UN number : 3439 Chem class : 1 Pack group : III


Quantity 25g

 View more versions of this product

Product Code. 11905641

  • £144.00 / 25g


Estimated Shipment
08-02-2018
Add to basket


Specifications

Chemical Identifiers

5-Amino-2-methylbenzonitrile
96.0
≥96.0%
C8H8N2
50670-64-9
100.0
97%
132.17

Specifications

5-Amino-2-methylbenzonitrile
Authentic
100.0
Yellow to brown
≥96.0%
97%
132.17
50670-64-9
96.0
88° to 93°C
Powder
25g
C8H8N2
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 5-Amino-2-methylbenzonitrile, 97%

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4

Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ A§ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/A§.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS