missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Amino-2-fluorophenol, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
5-Amino-2-fluorophenol is used as synthetic building block.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
5-Amino-2-fluorophenol is used as synthetic building block.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Specifications
Melting Point | 156°C to 158°C |
Color | White to Yellow |
Quantity | 1 g |
Formula Weight | 127.12 |
Percent Purity | 97% |
Physical Form | Solid |
Chemical Name or Material | 5-Amino-2-fluorophenol |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'