missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2-fluoro-4-methylbenzene boronic acid pinacol ester, 96%, Thermo Scientific™

Catalog Number p-7054304
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1817801-55-0
C13H19BFNO2
251.11
MONFQILBYDTFKM-UHFFFAOYSA-N
4-fluoro-2-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline
This item is not returnable. View return policy
1817801-55-0
C13H19BFNO2
251.11
MONFQILBYDTFKM-UHFFFAOYSA-N
4-fluoro-2-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 251.1
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 5-Amino-2-fluoro-4-methylbenzene boronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'