missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Amino-2-bromo-6-methylpyridine, 98%

Brand:  Alfa Aesar™ H64756.03

Code : NEW

Additional Details : CAS Number : 126325-47-1 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15402089

  • £10.05 / 1g

Estimated Shipment: 26-08-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

126325-47-1
187.04
UBTQJSZELCWGCN-UHFFFAOYSA-N
14418045
CC1=C(N)C=CC(Br)=N1
C6H7BrN2
MFCD03095086
5-amino-2-bromo-6-picoline, 3-amino-6-bromo-2-methylpyridine, 2-bromo-5-amino-6-methylpyridine, 3-amino-6-bromo-2-picoline, 5-amino-2-bromo-6-methylpyridine, 2-bromo-5-amino-6-picoline, 6-bromo-2-methyl-3-pyridylamine, 6-bromo-2-methyl-pyridin-3-ylamine, 2-methyl-3-amino-6-bromopyridine, pubchem1117
6-bromo-2-methylpyridin-3-amine

Specifications

5-Amino-2-bromo-6-methylpyridine
110°C to 111°C
126325-47-1
MFCD03095086
5-amino-2-bromo-6-picoline, 3-amino-6-bromo-2-methylpyridine, 2-bromo-5-amino-6-methylpyridine, 3-amino-6-bromo-2-picoline, 5-amino-2-bromo-6-methylpyridine, 2-bromo-5-amino-6-picoline, 6-bromo-2-methyl-3-pyridylamine, 6-bromo-2-methyl-pyridin-3-ylamine, 2-methyl-3-amino-6-bromopyridine, pubchem1117
CC1=C(N)C=CC(Br)=N1
187.04
187.04
98%
White
1g
C6H7BrN2
6190039
UBTQJSZELCWGCN-UHFFFAOYSA-N
6-bromo-2-methylpyridin-3-amine
14418045
Solid
Videos
SDS Documents
Certificates