missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Amino-2-bromo-6-methylpyridine, 98%

Brand:  Alfa Aesar™ H64756.06

Code : NEW

Additional Details : CAS Number : 126325-47-1 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15412089

  • POA / 5g

This item cannot be ordered online because the price is subject to change. Please contact our Product Support team on 01509 555500 for assistance.


Specifications

Chemical Identifiers

5-Amino-2-bromo-6-methylpyridine
187.04
6190039
126325-47-1
MFCD03095086
5-Amino-2-bromo-6-picoline

Specifications

5-Amino-2-bromo-6-methylpyridine
Solid
110° to 111°C
187.04
MFCD03095086
5-Amino-2-bromo-6-picoline
126325-47-1
98%
Off-white
5g
6190039
Videos
SDS