missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Amino-2,4-dichlorophenol, 97%

Brand:  Alfa Aesar™ H66753.03

Code : NEW

Additional Details : CAS Number : 39489-79-7 Weight : 0.00100kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III

Quantity 1g

 View more versions of this product

Product Code. 15442819

  • £27.10 / 1g


Estimated Shipment date
from Supplier 25-05-2018
Add to basket

Specifications

Chemical Identifiers

5-Amino-2,4-dichlorophenol
178.01
UN2811
2,4-Dichloro-5-hydroxyaniline
39489-79-7
MFCD00186187
2717129

Specifications

5-Amino-2,4-dichlorophenol
133° to 137°C
97%
178.01
UN2811
2,4-Dichloro-5-hydroxyaniline
39489-79-7
Solid
1g
MFCD00186187
2717129
Videos
SDS