missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-1H-benzimidazole-2-thiol, 97%, Thermo Scientific™

Catalog Number p-4813181
Click to view available options
Quantity:
1 g
10 g
50 g
This item is not returnable. View return policy
2818-66-8
C7H7N3S
165.21
MFCD00022671
BXDMTLVCACMNJO-UHFFFAOYSA-N
5-amino-2-mercaptobenzimidazole, 5-amino-2-benzimidazolethiol, 5-amino-1h-benzo d imidazole-2-thiol, 5-amino-1h-benzimidazole-2-thiol, 5-aminobenzimidazole-2-thiol, 2h-benzimidazole-2-thione, 5-amino-1,3-dihydro, 2-benzimidazolethiol, 5-amino, 5-amino-1h-1,3-benzodiazole-2-thiol, 2-mercapto-5-amino benzimidazole, 5-amino-3h-1,3-benzodiazole-2-thiol
2734002
NC1=CC=C2NC(=S)NC2=C1
This item is not returnable. View return policy
2818-66-8
C7H7N3S
165.21
MFCD00022671
BXDMTLVCACMNJO-UHFFFAOYSA-N
5-amino-2-mercaptobenzimidazole, 5-amino-2-benzimidazolethiol, 5-amino-1h-benzo d imidazole-2-thiol, 5-amino-1h-benzimidazole-2-thiol, 5-aminobenzimidazole-2-thiol, 2h-benzimidazole-2-thione, 5-amino-1,3-dihydro, 2-benzimidazolethiol, 5-amino, 5-amino-1h-1,3-benzodiazole-2-thiol, 2-mercapto-5-amino benzimidazole, 5-amino-3h-1,3-benzodiazole-2-thiol
2734002
NC1=CC=C2NC(=S)NC2=C1

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Assay Percent Range 97%
Quantity 1 g
IUPAC Name 5-amino-1,3-dihydrobenzimidazole-2-thione
Formula Weight 165.22
Chemical Name or Material 5-Amino-1H-benzimidazole-2-thiol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'