missing translation for 'onlineSavingsMsg'
Learn More

5-Acetylvaleric acid, 90+%, technical, Thermo Scientific Chemicals

Catalog Number 13488668
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
3128-07-2
C7H10O3
144.17
IZOQMUVIDMLRDC-UHFFFAOYSA-N
5-acetylvaleric acid, heptanoic acid, 6-oxo, 5-aceto valeric acid, 6-oxo-heptanoic acid, unii-4u902i8qzo, 5-acetylvalericacid, 6-oxoenanthic acid, 6-ketoheptanoic acid, 5-acetylpentanoic acid, #
98810
6-oxoheptanoic acid
CC(=O)CCCCC(=O)O
This item is not returnable. View return policy
3128-07-2
C7H10O3
144.17
IZOQMUVIDMLRDC-UHFFFAOYSA-N
5-acetylvaleric acid, heptanoic acid, 6-oxo, 5-aceto valeric acid, 6-oxo-heptanoic acid, unii-4u902i8qzo, 5-acetylvalericacid, 6-oxoenanthic acid, 6-ketoheptanoic acid, 5-acetylpentanoic acid, #
98810
6-oxoheptanoic acid
CC(=O)CCCCC(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 35°C to 37°C
Boiling Point 138°C to 140°C (1.5 mmHg)
Assay Percent Range 90% min. (GC)
Packaging Glass bottle
Linear Formula CH3C(O)(CH2)4CO2H
Quantity 25 g
Solubility Information Solubility in water: soluble
Formula Weight 144.17
Percent Purity 90+%
Grade Technical
Chemical Name or Material 5-Acetylvaleric acid
Show More Show Less
compliance-icons
Product Identifier
  • 5-Acetylvaleric acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'