missing translation for 'onlineSavingsMsg'
Learn More

5-Acetyl-2-thiophenecarbaldehyde, 97%, Maybridge™

Brand:  Maybridge MO01240DE

Code : 31

Additional Details : CAS Number : 4565-29-1 Weight : 0.05500kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III

Quantity 5g

 View more versions of this product

Product Code. 10002314

  • £322.00 / 5g

Estimated Shipment: 03-09-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

4565-29-1
154.183
5-acetyl-2-thiophenecarbaldehyde, 2-acetyl-5-formylthiophene, 5-acetyl-thiophene-2-carbaldehyde, 2-thiophenecarboxaldehyde,5-acetyl
5-acetylthiophene-2-carbaldehyde
C7H6O2S
ISGDPVVLVWQENT-UHFFFAOYSA-N
2794713
CC(=O)C1=CC=C(S1)C=O

Specifications

5-Acetyl-2-thiophenecarbaldehyde
5g
97%
5-acetyl-2-thiophenecarbaldehyde, 2-acetyl-5-formylthiophene, 5-acetyl-thiophene-2-carbaldehyde, 2-thiophenecarboxaldehyde,5-acetyl
CC(=O)C1=CC=C(S1)C=O
154.183
154.19
Amber Glass Bottle
4565-29-1
C7H6O2S
ISGDPVVLVWQENT-UHFFFAOYSA-N
5-acetylthiophene-2-carbaldehyde
2794713
97%
Videos
SDS Documents
Certificates