missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Acetyl-2-hydroxybenzamide, 97%, Thermo Scientific™
Click to view available options
Quantity:
50 g
100 g
Specifications
Assay Percent Range | 97% |
Quantity | 50 g |
Formula Weight | 179.17 |
Chemical Name or Material | 5-Acetyl-2-hydroxybenzamide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'