Learn More
Tocris Bioscience™ 5,7-Dichlorokynurenic acid sodium salt
Sodium salt of 5,7-Dichlorokynurenic acid (Cat. No. 0286). Potent NMDA antagonist, acts at glycine site
Brand:  Tocris Bioscience™ 369850
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13433444
  • £398.37 / 50mg
Estimated Shipment date
from Supplier 02-04-2020
Description
Description
Sodium salt of 5,7-Dichlorokynurenic acid (Cat. No. 0286), a potent antagonist at the glycine site of the NMDA receptor (Ki = 79 nM vs. [3H]-glycine).
Specifications
Specifications
5,7-Dichlorokynurenic acid sodium salt
>98
50mg
5,7-Dichlorokynurenic acid sodium salt
280.04