missing translation for 'onlineSavingsMsg'
Learn More

5,7-Dichloro-8-hydroxyquinoline, 98%, Thermo Scientific Chemicals

Catalog Number p-105305
Click to view available options
Quantity:
5 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
773-76-2
C9H5Cl2NO
214.05
MFCD00006786
WDFKMLRRRCGAKS-UHFFFAOYSA-N
chloroxine, 5,7-dichloro-8-hydroxyquinoline, chlorquinol, capitrol, dichloroxin, quixalin, chloroxyquinoline, dichloroquinolinol, dikhloroskin, 5,7-dichloro-8-quinolinol
2722
CHEBI:59477
5,7-dichloroquinolin-8-ol
C1=CC2=C(C(=C(C=C2Cl)Cl)O)N=C1
This item is not returnable. View return policy
773-76-2
C9H5Cl2NO
214.05
MFCD00006786
WDFKMLRRRCGAKS-UHFFFAOYSA-N
chloroxine, 5,7-dichloro-8-hydroxyquinoline, chlorquinol, capitrol, dichloroxin, quixalin, chloroxyquinoline, dichloroquinolinol, dikhloroskin, 5,7-dichloro-8-quinolinol
2722
CHEBI:59477
5,7-dichloroquinolin-8-ol
C1=CC2=C(C(=C(C=C2Cl)Cl)O)N=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 178°C to 183°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Beilstein 21, 95
Merck Index 15, 2181
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in water at ph 1 or >9;,slightly soluble in ethanol,acetone,chloroform,,and dmso;,soluble in alkali,benzene,petroleum ether
Formula Weight 214.05
Percent Purity 98%
Physical Form Fluffy Powder
Chemical Name or Material 5, 7-Dichloro-8-hydroxyquinoline, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 5,7-Dichloro-8-hydroxyquinoline
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'