missing translation for 'onlineSavingsMsg'
Learn More

5,6-Dimethoxyindole, 99%, Thermo Scientific™

Catalog Number p-3778352
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
14430-23-0
C10H11NO2
177.20
MFCD00005675
QODBZRNBPUPLEZ-UHFFFAOYSA-N
84431
5,6-dimethoxy-1H-indole
COC1=C(OC)C=C2C=CNC2=C1
This item is not returnable. View return policy
14430-23-0
C10H11NO2
177.20
MFCD00005675
QODBZRNBPUPLEZ-UHFFFAOYSA-N
84431
5,6-dimethoxy-1H-indole
COC1=C(OC)C=C2C=CNC2=C1
CAS Min % 98.5
CAS Max % 100.0
Melting Point 154.0°C
Color Beige
Boiling Point 198.0°C (8.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Synonym 5,6-dimethoxyindole, 1h-indole, 5,6-dimethoxy, 5,6-dimethoxyindol, zlchem 178, pubchem7329, acmc-1cbhq, 1h-indole,5,6-dimethoxy, #
Solubility Information Solubility in water: insoluble
Formula Weight 177.2
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 5, 6-Dimethoxyindole
Show More Show Less
compliance-icons
Product Identifier
  • 5,6-Dimethoxyindole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'