missing translation for 'onlineSavingsMsg'
Learn More

5,6-Dichloropyridine-2-carboxylic acid, 98%, Thermo Scientific Chemicals

Catalog Number 15401479
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
88912-24-7
C6H3Cl2NO2
191.995
MFCD13185546
QOJNAEPFSUYAFL-UHFFFAOYSA-N
5,6-dichloropicolinic acid, 5,6-dichloro-2-pyridinecarboxylic acid, 2-pyridinecarboxylic acid, 5,6-dichloro, 2,3-dichloro-6-carboxypyridine, 2-pyridinecarboxylicacid, 5,6-dichloro, acmc-209vdz, 2,3-dichloro-6-pyridinecarboxylicacid, 5,6-dichloro-pyridine-2-carboxylic acid
13278016
5,6-dichloropyridine-2-carboxylic acid
C1=CC(=NC(=C1Cl)Cl)C(=O)O
This item is not returnable. View return policy
88912-24-7
C6H3Cl2NO2
191.995
MFCD13185546
QOJNAEPFSUYAFL-UHFFFAOYSA-N
5,6-dichloropicolinic acid, 5,6-dichloro-2-pyridinecarboxylic acid, 2-pyridinecarboxylic acid, 5,6-dichloro, 2,3-dichloro-6-carboxypyridine, 2-pyridinecarboxylicacid, 5,6-dichloro, acmc-209vdz, 2,3-dichloro-6-pyridinecarboxylicacid, 5,6-dichloro-pyridine-2-carboxylic acid
13278016
5,6-dichloropyridine-2-carboxylic acid
C1=CC(=NC(=C1Cl)Cl)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Beilstein 4390497
Formula Weight 192
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 5,6-Dichloropyridine-2-carboxylic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'