missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5,6-Dichloro-1H-benzo[d]imidazole-2-thiol, ≥95%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Specifications
CAS Min % | 94.5 |
CAS Max % | 100.0 |
Assay Percent Range | 95% |
Quantity | 5 g |
Formula Weight | 219.09 |
Percent Purity | 95% |
Chemical Name or Material | 5, 6-Dichloro-1H-benzo[d]imidazole-2-thiol |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'