missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5,6,7,8-Tetrahydro-9H-pyrido[3,2-b]indol-9-one, 98%

Brand:  Alfa Aesar™ H35275.MD

Code : NEW

Additional Details : CAS Number : 198141-12-7 Weight : 0.00025kg

Quantity 250mg

 View more versions of this product

Product Code. 15488995

  • £133.80 / 250mg


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

5,6,7,8-Tetrahydro-9H-pyrido[3,2-b]indol-9-one
198141-12-7
186.21

Specifications

5,6,7,8-Tetrahydro-9H-pyrido[3,2-b]indol-9-one
265°C (dec.)
198141-12-7
98%
Pale brown
250mg
Crystalline Powder
186.21
Videos
SDS