missing translation for 'onlineSavingsMsg'
Learn More

5,6,7,8-Tetrahydro-2-naphthol, 98%, Thermo Scientific Chemicals

Catalog Number p-3830118
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1125-78-6
C10H12O
148.2
MFCD00001738
UMKXSOXZAXIOPJ-UHFFFAOYSA-N
5,6,7,8-tetrahydro-2-naphthol, 6-tetralinol, 6-hydroxytetralin, 2-naphthalenol, 5,6,7,8-tetrahydro, 5,6,7,8-tetrahydro-beta-naphthol, ac-beta-tetralol, 5,6,7,8-tetrahydro-2-hydroxynaphthalene, 2-naphthol, 5,6,7,8-tetrahydro, unii-tmr02i7n8s, 5,6,7,8-tetrahydro-2-naphthalenol
14305
CHEBI:34448
5,6,7,8-tetrahydronaphthalen-2-ol
C1CCC2=C(C1)C=CC(=C2)O
This item is not returnable. View return policy
1125-78-6
C10H12O
148.2
MFCD00001738
UMKXSOXZAXIOPJ-UHFFFAOYSA-N
5,6,7,8-tetrahydro-2-naphthol, 6-tetralinol, 6-hydroxytetralin, 2-naphthalenol, 5,6,7,8-tetrahydro, 5,6,7,8-tetrahydro-beta-naphthol, ac-beta-tetralol, 5,6,7,8-tetrahydro-2-hydroxynaphthalene, 2-naphthol, 5,6,7,8-tetrahydro, unii-tmr02i7n8s, 5,6,7,8-tetrahydro-2-naphthalenol
14305
CHEBI:34448
5,6,7,8-tetrahydronaphthalen-2-ol
C1CCC2=C(C1)C=CC(=C2)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 58°C to 62°C
Color Beige
Boiling Point 275°C to 276°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 10 g
Beilstein 06, 579
Formula Weight 148.2
Percent Purity 98%
Physical Form Crystalline Mass
Chemical Name or Material 5,6,7,8-Tetrahydro-2-naphthol, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 5,6,7,8-Tetrahydro-2-naphthol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'