missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-[5-(Trifluoromethyl)isoxazol-3-yl]thiophene-2-sulfonyl chloride, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
10 g
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Packaging | Amber glass bottle |
Quantity | 250 mg |
Formula Weight | 317.69 |
Percent Purity | 97% |
Chemical Name or Material | 5-[5-(Trifluoromethyl)isoxazol-3-yl]thiophene-2-sulfonyl chloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'