missing translation for 'onlineSavingsMsg'
Learn More

5,5-Diphenylhydantoin, 99+%, ACROS Organics™

Brand:  Acros Organics 171735000

Code : 29

Additional Details : CAS Number : 57-41-0 Weight : 0.55000kg


Quantity 500g
Packaging Plastic bottle

 View more versions of this product

Product Code. 10676572

  • £116.60 / 500g


Estimated Shipment
20-06-2018
Add to basket


Specifications

Chemical Identifiers

5, 5-Diphenylhydantoin
57-41-0
100.0
99+%
252.27
Phenytoin
99+%
99.0
≥99.0% (on dry substance)
C15H12N2O2
MFCD00005264

Specifications

5, 5-Diphenylhydantoin
57-41-0
100.0
0.5% max. (1 g, 105°C)
Powder
99+%
252.27
24, 410
Phenytoin
500g
99+%
99.0
Authentic
Plastic bottle
≥99.0% (on dry substance)
C15H12N2O2
MFCD00005264
15, 7433
White to almost white
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 5,5-Diphenylhydantoin

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Reproductive toxicity Category 2

Hazard Statement
  • H351-Suspected of causing cancer.
  • H361d-Suspected of damaging the unborn child.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

SDS