missing translation for 'onlineSavingsMsg'
Learn More

5-(4-Pyridyl)-1,3,4-oxadiazole-2-thiol, 97%, Thermo Scientific™

Catalog Number p-7031730
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
15264-63-8
C7H5N3OS
179.20
MFCD00215200
TXCXZVFDWQYTIC-UHFFFAOYSA-N
5-4-pyridyl-1,3,4-oxadiazole-2-thiol, 5-pyridin-4-yl-1,3,4-oxadiazole-2-thiol, 5-pyridin-4-yl-1,3,4 oxadiazole-2-thiol, 5-pyridin-4-yl-1,3,4-oxadiazole-2 3h-thione, 5-4-pyridinyl-1,3,4-oxadiazole-2-thiol, 1,3,4-oxadiazole-2-thiol, 5-4-pyridinyl, 1,3,4-oxadiazole-2 3h-thione,5-4-pyridinyl, 5-pyridin-4-yl-3h-1,3,4-oxadiazole-2-thione, d04qyn
774383
S=C1NN=C(O1)C1=CC=NC=C1
This item is not returnable. View return policy
15264-63-8
C7H5N3OS
179.20
MFCD00215200
TXCXZVFDWQYTIC-UHFFFAOYSA-N
5-4-pyridyl-1,3,4-oxadiazole-2-thiol, 5-pyridin-4-yl-1,3,4-oxadiazole-2-thiol, 5-pyridin-4-yl-1,3,4 oxadiazole-2-thiol, 5-pyridin-4-yl-1,3,4-oxadiazole-2 3h-thione, 5-4-pyridinyl-1,3,4-oxadiazole-2-thiol, 1,3,4-oxadiazole-2-thiol, 5-4-pyridinyl, 1,3,4-oxadiazole-2 3h-thione,5-4-pyridinyl, 5-pyridin-4-yl-3h-1,3,4-oxadiazole-2-thione, d04qyn
774383
S=C1NN=C(O1)C1=CC=NC=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 262°C to 264°C
Quantity 1 g
IUPAC Name 5-pyridin-4-yl-3H-1,3,4-oxadiazole-2-thione
Formula Weight 179.2
Percent Purity 97%
Chemical Name or Material 5-(4-Pyridyl)-1,3,4-oxadiazole-2-thiol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'