missing translation for 'onlineSavingsMsg'
Learn More

5-(4-Nitrophenyl)furfural, 98%, Acros Organics

Brand:  Acros Organics 372730050

Code : 29

Additional Details : CAS Number : 7147-77-5 Weight : 0.05500kg

Quantity 5g

 View more versions of this product

Product Code. 10406953

  • £44.50 / 5g


Estimated Shipment
14-02-2018
Add to basket

Specifications

Chemical Identifiers

5-(4-Nitrophenyl)furfural
C11H7NO4
5-(4-Nitrophenyl)-2-furancarboxaldehyde
98%
217.18

Specifications

5-(4-Nitrophenyl)furfural
Glass bottle
C11H7NO4
5g
7147-77-5
98%
217.18
5-(4-Nitrophenyl)-2-furancarboxaldehyde
Videos
SDS