missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-(4-Nitrophenyl)-1H-tetrazole, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
It is used as a reagent used as activator for both solution- and solid-phase synthesis of oligonucleotides using the phosphoramidite method.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as a reagent used as activator for both solution- and solid-phase synthesis of oligonucleotides using the phosphoramidite method.
Solubility
Insoluble in water. Soluble in DMSO.
Notes
It may be stored in the dark in a desiccator. Avoid contact with strong heat and metal salts.
It is used as a reagent used as activator for both solution- and solid-phase synthesis of oligonucleotides using the phosphoramidite method.
Solubility
Insoluble in water. Soluble in DMSO.
Notes
It may be stored in the dark in a desiccator. Avoid contact with strong heat and metal salts.
Specifications
Melting Point | ∽223°C (decomposition) |
Quantity | 1 g |
Beilstein | 192840 |
Solubility Information | Insoluble in water. Soluble in DMSO. |
Formula Weight | 191.15 |
Percent Purity | 97% |
Chemical Name or Material | 5-(4-Nitrophenyl)-1H-tetrazole |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'