missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-(4-Iodophenyl)isoxazole, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber glass bottle |
Quantity | 250 mg |
Formula Weight | 271.05 |
Percent Purity | 97% |
Chemical Name or Material | 5-(4-Iodophenyl)isoxazole |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'