missing translation for 'onlineSavingsMsg'
Learn More

5-(4-Ethynylphenyl)-1,3-oxazole, 97%, Maybridge™

£50.00 - £129.00

Chemical Identifiers

CAS 501944-63-4
Molecular Formula C11H7NO
Molecular Weight (g/mol) 169.183
MDL Number MFCD08435847
InChI Key CYRYZDZQFDRTHD-UHFFFAOYSA-N
Synonym 5-4-ethynylphenyl-1,3-oxazole, 5-4-ethynylphenyl oxazole, oxazole,5-4-ethynylphenyl, 4-1,3-oxazol-5-yl phenylacetylene
PubChem CID 18525707
IUPAC Name 5-(4-ethynylphenyl)-1,3-oxazole
SMILES C#CC1=CC=C(C=C1)C2=CN=CO2
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10751464
SDS View Documents
Maybridge
CC14547CB
250mg Amber Glass Bottle
£50.00
250mg
View alternative products
Save Up to 
»
Estimated Shipment: 21-10-2021
Please to purchase this item. Need a web account? Register with us today!
10422254
SDS View Documents
Maybridge
CC14547DA
1g Amber Glass Bottle
£129.00
1g
View alternative products
Save Up to 
»
Estimated Shipment: 21-10-2021
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

501944-63-4
169.183
CYRYZDZQFDRTHD-UHFFFAOYSA-N
18525707
C#CC1=CC=C(C=C1)C2=CN=CO2
C11H7NO
MFCD08435847
5-4-ethynylphenyl-1,3-oxazole, 5-4-ethynylphenyl oxazole, oxazole,5-4-ethynylphenyl, 4-1,3-oxazol-5-yl phenylacetylene
5-(4-ethynylphenyl)-1,3-oxazole

Specifications

5-(4-Ethynylphenyl)-1, 3-oxazole
250mg
96.5
97%
MFCD08435847
CYRYZDZQFDRTHD-UHFFFAOYSA-N
5-(4-ethynylphenyl)-1,3-oxazole
18525707
97%
Amber Glass Bottle
501944-63-4
100.0
C11H7NO
5-4-ethynylphenyl-1,3-oxazole, 5-4-ethynylphenyl oxazole, oxazole,5-4-ethynylphenyl, 4-1,3-oxazol-5-yl phenylacetylene
C#CC1=CC=C(C=C1)C2=CN=CO2
169.183
169.18
SDS
Product Certifications