missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 5-(4-Dimethylaminobenzylidene)rhodanine, 99%

Catalog Number p-115801
29.53 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
536-17-4
C12H12N2OS2
264.35
MFCD00064857
JJRVRELEASDUMY-UHFFFAOYSA-N
unii-0ser53q7rt, p-dimethylaminobenzalrhodanine, 5-4-dimethylaminobenzylidene rhodanine, 0ser53q7rt, usaf pd-20, 4-dimethylaminobenzylidenerhodanine, 4-thiazolidinone, 5-4-dimethylamino phenyl methylene-2-thioxo, 5-p-dimethylaminobenzal rhodanine, p-dimethylaminobenzylidene rhodamine, p-dimethylamino benzal-5-rhodanine
2723826
5-[[4-(dimethylamino)phenyl]methylidene]-2-sulfanylidene-1,3-thiazolidin-4-one
CN(C)C1=CC=C(C=C1)C=C2C(=O)NC(=S)S2
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
536-17-4
C12H12N2OS2
264.35
MFCD00064857
JJRVRELEASDUMY-UHFFFAOYSA-N
unii-0ser53q7rt, p-dimethylaminobenzalrhodanine, 5-4-dimethylaminobenzylidene rhodanine, 0ser53q7rt, usaf pd-20, 4-dimethylaminobenzylidenerhodanine, 4-thiazolidinone, 5-4-dimethylamino phenyl methylene-2-thioxo, 5-p-dimethylaminobenzal rhodanine, p-dimethylaminobenzylidene rhodamine, p-dimethylamino benzal-5-rhodanine
2723826
5-[[4-(dimethylamino)phenyl]methylidene]-2-sulfanylidene-1,3-thiazolidin-4-one
CN(C)C1=CC=C(C=C1)C=C2C(=O)NC(=S)S2

5-(4-Dimethylaminobenzylidene)rhodanine, 99%, ACROS Organics, Quantity: 100g, Packaging: Glass bottle, Red, Melting Point: 285 deg.C to 288 deg.C, Molecular Weight: 264.35, Percent Purity: 99%, Appearance: (0.3% soln. in acetone) clear, Assay Percent Range: 99%, Beilstein: 27, 433, CAS: 536-17-4

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 285°C to 288°C
Color Red
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 10 g
Beilstein 27, 433
Merck Index 15, 3253
Solubility Information (0.03% in acetone) Clear
Formula Weight 264.35
Percent Purity 99%
Physical Form Fluffy Powder
Chemical Name or Material 5-(4-Dimethylaminobenzylidene)rhodanine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'